Learn More
VITLAB™ Polypropylene Y-shape Connectors
Brand:  VITLAB™ 80465
Additional Details : Weight : 0.00400kg
 View more versions of this product
Product Code. 15272759
  • 11.91 SEK / Pack of 20
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Specifications
Specifications
Yes
Connector
20 Pack
Polypropylene
For 4 to 5mm tubing I.D.
Y-shape
Videos