Läs mer
Polyquaternium 11 (20% in H2O), TRC
Produktkod. 30421600
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30421600 10 g
1 options
Produktkod. 30421600 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP68813010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Polyquaternium 11
CAS 53633-54-8
Molekylformel C18H35N2O3•C2H5O4S•(C2H4)x•(C3H6)y
Kvantitet 10 g
Synonym 1-Ethenyl-2-pyrrolidinone Polymer With 2-(Dimethylamino)ethyl 2-Methyl-2-propenoate Compound With Diethyl Sulfate, Sulfuric Acid Diethyl Ester Compound With 2-(Dimethylamino)ethyl 2-Methyl-2-propenoate Polymer With 1-Ethenyl-2-pyrrolidinone, Celquat 200, Copolymer 755, Dehyquart CC 11, Flocare C 111, Gafquat 440, Gafquat 734, Gafquat 755, Gafquat 755N, Gafquat 755N-P, Gafquat 755N-PP, Gafquat 755N-PW, HC 1NS, HC Polymer 1N, HC Polymer 1NS, HC Polymer 1S, HC Polymer 1S(M), HC Polymer 2L, HC Polymer 3A, HC Polymer 5, Luviquat PQ 11, Luviquat PQ 11PN, N,N-Dimethylaminoethyl Methacrylate-vinylpyrrolidone Copolymer Diethyl Sulfate Salt, PQ 11, PQ 11PN, Polyquat 11, Polyquaternium 11, Quaternium 23
InChI formel InChI=1S/C8H15NO2.C6H9NO.C4H10O4S/c1-7(2)8(10)11-6-5-9(3)4;1-2-7-5-3-4-6(7)8;1-3-7-9(5,6)8-4-2/h1,5-6H2,2-4H3;2H,1,3-5H2;3-4H2,1-2H3
Molekylvikt (g/mol) 327.4812512281
Koncentration eller sammansättning (efter analyt eller komponenter) 20 %