Läs mer
Polystyrene crosslinked with divinylbenzene, brominated, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Polystyrene crosslinked with divinylbenzene, brominated |
| CAS | 88497-56-7 |
| Molekylformel | [CH2CH(C6H4)n(Br)n |
| Kvantitet | 100 mg |
| Synonym | Brominated Polystyrene, Copolymer of styrene and divinylbenzene, brominated, Polystyrene crosslinked with divinylbenzene, brominated. |
| InChI formel | InChI=1S/C10H13Br/c1-3-8(2)9-4-6-10(11)7-5-9/h4-8H,3H2,1-2H3 |
| LEDER | CCC(C)c1ccc(Br)cc1 |
| IUPAC-namn | 1-bromo-4-butan-2-ylbenzene |
| Molekylvikt (g/mol) | 213.11 |
| Formel vikt | 212.0201 |