Learn More
Alfa Aesar™ Polystyrene standard, M.W. 400,000, Mw/Mn 1.06
Brand:  Alfa Aesar™ 041939.03
UNSPSC : 13102026
Additional Details : CAS Number : 9003-53-6 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15461747
  • 648.50 SEK / 1g
Estimated Shipment date
from Supplier 04-09-2017
Add to basket
Specifications
Chemical Identifiers
Polystyrene standard
9003-53-6
M.W. 400,000, Mw/Mn 1.06
MFCD00084450
Specifications
Polystyrene standard
9003-53-6
MFCD00084450
1g
M.W. 400,000, Mw/Mn 1.06
Powder
Odorless
Documentation
Technical Documents