missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
TdB Labs AB Polysucrose for Cell Separation and Molecular Filtration
Beskrivning
- Product Overview: Polysucrose 1000 kDa is a neutral, highly branched polysaccharide used for cellular and molecular separations. With high water solubility and biocompatibility, it is widely applied in gradient separation, size exclusion chromatography, and barrier diffusion studies.
- Features:
- High Molecular Weight: Approximately 1000 kDa for modeling large macromolecule behavior.
- Water-Soluble and Inert: Provides minimal interaction with biological components.
- Neutral Charge: Reduces nonspecific binding during separation or diffusion processes.
- Applications:
- Cell separation by density gradients.
- Molecular sieving and size exclusion studies.
- Modeling extracellular matrix-like diffusion behaviors.
- Standard for calibrating large molecule diffusion assays.
Specifikationer
Specifikationer
| Kvantitet | 1 g |
| Molekylvikt (g/mol) | 50 kDa |
| Kvalitet | Research |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?