missing translation for 'onlineSavingsMsg'
Läs mer

TdB Labs AB Polysucrose for SEC-chromatography and Cell Sorting in Biomedical Studies

Produktkod. 30216442
Encompass_Preferred
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
Förpackningsstorlek:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
30216442 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30216442 Leverantör TdB Labs AB Leverantörsnummer P40 _1g
 Begränsad artikel
Beräknad leverans: 26-11-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Polysucrose 20 kDa is a low molecular weight, neutral branched polysaccharide used in tracer diffusion, permeability, and extracellular volume studies. Its compact structure offers high solubility and compatibility with biological systems.

  • Product Overview: Polysucrose 1000 kDa is a neutral, highly branched polysaccharide used for cellular and molecular separations. With high water solubility and biocompatibility, it is widely applied in gradient separation, size exclusion chromatography, and barrier diffusion studies.
  • Features:
    • High Molecular Weight: Approximately 1000 kDa for modeling large macromolecule behavior.
    • Water-Soluble and Inert: Provides minimal interaction with biological components.
    • Neutral Charge: Reduces nonspecific binding during separation or diffusion processes.
  • Applications:
    • Cell separation by density gradients.
    • Molecular sieving and size exclusion studies.
    • Modeling extracellular matrix-like diffusion behaviors.
    • Standard for calibrating large molecule diffusion assays.

Specifikationer

Kvantitet 1 g
Molekylvikt (g/mol) 40 kDa
Kvalitet Research
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.