Learn More
Ark Plas Products™ Polyurethane Tubing
An excellent choice for tubing in the laboratory, offering excellent resistance to chemicals, kinking, abrasion, oxidation and hydrolysis.
Brand:  Ark Plas Products™ TAX79-AN4
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11793159
  • 8403.06 SEK / Each
Estimated Shipment
08-07-2021
Add to basket
Description
Description
  • Provides flexibility and high impact strength for durability
  • Above average performance in cold temperatures
  • Extruded using state of the art inline process monitoring
  • Shipped bulk on cardboard spools
Specifications
Specifications
Blue
Autoclavable
Polyurethane
300 m
1 Pack
85 (Shore A)
9.6 mm
12.8 mm
1.58 mm