Läs mer
Polyvinyl Chloride, TRC
Produktkod. 17979454
Change view
Kvantitet:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17979454 2 mg
17918524 10 mg
17937544 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17979454 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP6993302MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 9002-86-2
Molekylformel (C2H3Cl)n
Molekylvikt (g/mol) 62.5
Synonym Ethylene, chloro-, polymers (8CI), 00178M7, 05NA, 080S, 1000Z, 101EP, 1032X, 103EP, 103EP8, 103EP8D, 103ZP, 108EP, 110A, 1170M55, 1185PC, 1233E, 1300H, 150-255 Natural, 168PE, 1800S, 195J, 1HH5NL
LEDER C=CCl
Specifikationer
Kemiskt namn eller material Poly(vinyl chloride)
Kvantitet 2 mg
InChI formel InChI=1S/C2H3Cl/c1-2-3/h2H,1H2
Formel vikt 61.9923