Learn More
Kautex™ PVC Wide Neck Square Container, Transparent, Without Closure
Fill and clean these wide neck PVC containers with ease due to the roomy neck configuration. Supplied without closure.
3.33 SEK - 4509.60 SEK
Specifications
Product Type | Wide Neck Container |
---|---|
Color | Transparent |
For Use With (Application) | Packaging, Dispensing, Sending Samples, Storage, Mailing |
Material | PVC |
Closure Included | No |
Product Code | Brand | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
17868167
|
Kautex™
2000084419 |
100 mL | 83.5 mm |
3759.70 SEK
Pack of 480 |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15941401
|
Kautex™
2000070596 |
100 mL | 71 mm |
626.59 SEK
Pack of 80 |
In Stock
Log in to see stock. |
||||
17858167
|
Kautex™
2000084412 |
200 mL | 98.5 mm |
3781.59 SEK
Pack of 288 |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10300771
|
Kautex™
2000070597 |
200 mL | 87 mm |
13.75 SEK
Each |
In Stock
Log in to see stock. |
||||
10050541
|
Kautex™
2000084423 |
300 mL | 105.5 mm |
3.33 SEK
Each |
In Stock
Log in to see stock. |
||||
17848167
|
Kautex™
2000084423 |
300 mL | 105.5 mm |
4509.60 SEK
Pack of 288 |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10733481
|
Kautex™
2000071080 |
300 mL | 94 mm |
18.45 SEK
Each |
In Stock
Log in to see stock. |
||||
17818167
|
Kautex™
2000084422 |
500 mL | 118.5 mm |
2399.88 SEK
Pack of 126 |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15971451
|
Kautex™
2000084422 |
500 mL | 118.5 mm |
84.75 SEK
Pack of 21 |
In Stock
Log in to see stock. |
||||
15951401
|
Kautex™
2000070598 |
500 mL | 108 mm |
3085.53 SEK
Pack of 162 |
In Stock
Log in to see stock. |
||||
15981411
|
Kautex™
2000071082 |
750 mL | 120 mm |
2711.94 SEK
Pack of 114 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
||||
10196730
|
Kautex™
2000070599 |
1000 mL | 143 mm |
24.95 SEK
Each |
In Stock
Log in to see stock. |
||||
15961401
|
Kautex™
2000070599 |
1000 mL | 143 mm |
2661.64 SEK
Pack of 96 |
In Stock
Log in to see stock. |
||||
17838127
|
Kautex™
2000073352 |
2000 mL | 187 mm |
2203.20 SEK
Pack of 44 |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10429860
|
Kautex™
2000070596 |
100 mL | 71 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10234491
|
Kautex™
2000084419 |
100 mL | 83.5 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10300721
|
Kautex™
2000070598 |
500 mL | 108 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10244491
|
Kautex™
2000084412 |
200 mL | 98.5 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10247830
|
Kautex™
2000071082 |
750 mL | 120 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15961451
|
Kautex™
2000084419 |
100 mL | 83.5 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10050551
|
Kautex™
2000084422 |
500 mL | 118.5 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10202091
|
Kautex™
2000073352 |
2000 mL | 187 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15951451
|
Kautex™
2000084412 |
200 mL | 98.5 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Member of Series 310 container family
- Closure available separately
Specifications
Wide Neck Container | |
Packaging, Dispensing, Sending Samples, Storage, Mailing | |
No |
Transparent | |
PVC | |
Square |