Läs mer
Tocris Bioscience™ Pomalidomide 4'-alkylC3-acid
Beskrivning
Pomalidomide 4'-alkylC3-acid is a functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand plus an alkylC3 linker with terminal acid ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D. PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 4-[[2-(2,6-Dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1 H-isoindol-4-yl]amino]butanoic acid |
| CAS | 2225940-47-4 |
| Kvantitet | 25 mg |
| Mål | Degrader Building Blocks |
| Molekylformel | C17H17N3O6 |
| Renhet | 0.95 |