Läs mer
Tocris Bioscience™ Pomalidomide 4'-PEG4-amine
Beskrivning
Pomalidomide 4'-PEG4-amine is a functionalized cereblon ligand for PROTACsup>® research and development; incorporates an E3 ligase ligand plus a PEG4 linker with terminal ethylamine ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
This product has been recently renamed. The previous name for this product was Pomalidomide - linker 8
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 4-[(14-Amino-3,6,9,12-tetraoxatetradec-1-yl)amino]-2-(2,6-dioxo-3-piperidinyl)-1H-Isoindole-1,3(2H)-dione hydrochloride |
| CAS | 2357105-92-9 |
| Synonym | Pomalidomide - linker 8 |
| Kvantitet | 25 mg |
| Molekylformel | C23H32N4O8.HCl |
| Formel vikt | 528.99 |
| Produkttyp | Ligand |
| Renhet | 95% |