Läs mer
MedChemExpress Pomalidomide-PEG3-C2-NH2 TFA
Produktkod. 16496016
Encompass_Preferred
Change view
Kvantitet:
100 mg
500 mg
1 g
2 g
Förpackningsstorlek:
500mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16496016 100 mg -
16491646 500 mg 500mg
16420186 2 g -
16481309 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16496016 Leverantör MedChemExpress Leverantörsnummer HY128716A
MedChemExpress Pomalidomide-PEG3-C2-NH2 TFA is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and 3-unit PEG linker used in PROTAC technology.
Specifikationer
Molekylvikt (g/mol) 562.49 g/mol
CAS 2414913-97-4
Innehåll och lagring 4°C, stored under nitrogen ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (stored under nitrogen)
Frakt skick Room temperature
Kvantitet 100 mg
Synonym Cereblon Ligand-Linker Conjugates 5 TFA ∣ E3 ligase Ligand-Linker Conjugates 30 TFA
Löslighet DMSO : 100 mg/mL (177.78 mM; Need ultrasonic) ∣ Ethanol : 100 mg/mL (177.78 mM; Need ultrasonic) ∣ H2O : 16.67 mg/mL (29.64 mM; Need ultrasonic)
Mål Dopamine Receptor