Learn More
Haldenwanger™ Porcelain Evaporating Basins
Glazed inside and out
Brand: Haldenwanger™ 130/3
Additional Details : Weight : 0.05000kg
Description
Deep form, French shape, glazed inside and out.Specifications
Round | |
Porcelain | |
Evaporating Basin | |
40 mm |
70 mL | |
Round | |
Glazed | |
75 mm |