missing translation for 'onlineSavingsMsg'
Learn More

Potassium 3-bromophenyltrifluoroborate, 97%, Thermo Scientific Chemicals

Catalog Number p-7047196
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
374564-34-8
C6H4BBrF3K
262.91
MFCD02093977
NIDYRVJZTADCGO-UHFFFAOYSA-N
potassium 3-bromophenyl trifluoroborate, potassium 3-bromophenyltrifluoroborate, potassium 3-bromophenyl trifluoroboranuide, amtb081, potassium 3-bromophenyl-trifluoroboranuide, potassium ion 3-bromophenyl trifluoroboranuide, potassium 3-bromophenyl trifluoro borate 1-, potassium 3-bromophenyl-tris fluoranyl boranuide
23677642
potassium;(3-bromophenyl)-trifluoroboranuide
[K+].F[B-](F)(F)C1=CC(Br)=CC=C1
This item is not returnable. View return policy
374564-34-8
C6H4BBrF3K
262.91
MFCD02093977
NIDYRVJZTADCGO-UHFFFAOYSA-N
potassium 3-bromophenyl trifluoroborate, potassium 3-bromophenyltrifluoroborate, potassium 3-bromophenyl trifluoroboranuide, amtb081, potassium 3-bromophenyl-trifluoroboranuide, potassium ion 3-bromophenyl trifluoroboranuide, potassium 3-bromophenyl trifluoro borate 1-, potassium 3-bromophenyl-tris fluoranyl boranuide
23677642
potassium;(3-bromophenyl)-trifluoroboranuide
[K+].F[B-](F)(F)C1=CC(Br)=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 178°C to 182°C
Quantity 1 g
Formula Weight 262.91
Percent Purity 97%
Chemical Name or Material Potassium 3-bromophenyltrifluoroborate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'