Läs mer
Potassium 4-Guaiacolsulfonate, TRC
Produktkod. 30419442
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30419442 50 mg
30415571 100 mg
30412852 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30419442 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP69500550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 16241-25-1
Molekylformel C7H7KO5S
Molekylvikt (g/mol) 242.29
Synonym 4-Hydroxy-3-methoxybenzenesulfonic Acid Potassium Salt (1:1), Potassium 4-hydroxy-3-methoxybenzenesulfonate
IUPAC-namn potassium;4-hydroxy-3-methoxybenzenesulfonate
LEDER [K+].COc1cc(ccc1O)S(=O)(=O)[O-]
Specifikationer
Kemiskt namn eller material Potassium 4-Guaiacolsulfonate
Kvantitet 50 mg
InChI formel InChI=1S/C7H8O5S.K/c1-12-7-4-5(13(9,10)11)2-3-6(7)8;/h2-4,8H,1H3,(H,9,10,11);/q;+1/p-1
Formel vikt 241.9651