missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Acetate, Granular, U.S.P., J.T. Baker™
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Specifications
Physical Form | Granules |
Quantity | 2.5 kg |
Formula Weight | 98.14 |
Grade | USP |
Packaging | Poly Bottle |
Specific Gravity | 1L = 1.57kg |
Chemical Name or Material | Potassium Acetate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'