missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromide, puriss p.a, ACS, ≥99.5% (AT), Honeywell Fluka™

Catalog Number p-7120685
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly bottle
7758-02-3
BrK
119.00
MFCD00011358
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
253877
CHEBI:32030
potassium;bromide
[K+].[Br-]
7758-02-3
BrK
119.00
MFCD00011358
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
253877
CHEBI:32030
potassium;bromide
[K+].[Br-]

Puriss. p.a., ACS Reagent,≥99.5% (AT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Boiling Point 1383°C
Melting Point 734°C
Color Colorless or White
pH 5.0 to 8.8
Quantity 250 g
Linear Formula KBr
UN Number NONH for all modes of transport
Formula Weight 119g/mol
Percent Purity ≥99.5% (AT)
Packaging Poly bottle
Vapor Pressure 1mmHg (795°C), 0.01mmHg (20°C)
Density 2.73g/mL
Chemical Name or Material Potassium bromide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'