Learn More
Potassium chloropalladite, 99.99%, BLD Pharm™
Product Code. 31029274
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Unit Size:
250mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31029274 25 g -
31022554 250 mg 250mg
31021204 5 g -
31029102 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31029274 Supplier BLD Pharm Supplier No. BD14891225g
 Restricted Item
Beräknad leverans: 27-11-2026
Specifications
CAS 10025-98-6
Quantity 25 g
Molecular Formula Cl4K2Pd
MDL Number MFCD00011373
SMILES Cl[Pd-2](Cl)(Cl)Cl.[K+].[K+]
Molecular Weight (g/mol) 326.43
Percent Purity 99.99%
Chemical Name or Material Potassium chloropalladite