Learn More
Potassium Citrate, Monohydrate, Granular, U.S.P. - F.C.C., J.T. Baker™
Catalog Number p-5390824
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Wide Mouth Amber Glass Bottle
6100-05-6
C6H7K3O8
324.41
PJAHUDTUZRZBKM-UHFFFAOYSA-K
potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate
2735208
CHEBI:64746
tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]
6100-05-6
C6H7K3O8
324.41
PJAHUDTUZRZBKM-UHFFFAOYSA-K
potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate
2735208
CHEBI:64746
tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]
Physical Form Granules
Quantity 500 g
Formula Weight 324.42
Grade FCC/USP
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 1.98kg
Chemical Name or Material Potassium Citrate, Monohydrate