Learn More
Potassium Cyanide, puriss. p.a. ACS Reagent, Solstice
Product Code. 15673330
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
25 kg
Unit Size:
100g
1kg
250g
25kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15673330 25 kg 25kg
15643330 100 g 100g
15663330 250 g 250g
15653330 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15673330 Supplier Honeywell-Fluka Supplier No. 3125225KG
  • ONLINE EXCLUSIVE PRICE
    74610.75 SEK
    54956.65 SEK / 25kg
    Save 19654.10 SEK
    26% Av
View all promotions
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥97.0%
  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 151-50-8
Molecular Formula CKN
Molecular Weight (g/mol) 65.116
MDL Number MFCD00011397
InChI Key NNFCIKHAZHQZJG-UHFFFAOYSA-N
Synonym potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
PubChem CID 9032
IUPAC Name potassium;cyanide
SMILES [C-]#N.[K+]
Specifications
Physical Form Solid
Quantity 25 kg
Linear Formula KCN
UN Number 1680
Beilstein 3593645
Formula Weight 65.12g/mol
Percent Purity ≥97%
Grade Not Available
Chemical Name or Material Potassium Cyanide, Reag. Ph. Eur.