Learn More
Potassium Cyanide, Supelco™
Specifications
Specifications
| Boiling Point | 1625°C/1013 hPa |
| Melting Point | 634°C |
| CAS | 151-50-8 |
| Quantity | 1 kg |
| MDL Number | MFCD00011397 |
| Synonym | Potassium cyanide, Cyanogen potassium, Potassium cyanide |
| Molecular Weight (g/mol) | 65.12 |
| Grade | Research |
| Density | 1.55 g/cm3 at 20°C |
| Chemical Name or Material | Potassium Cyanide |