Learn More
Potassium dichromate, MilliporeSigma™
Product Code. 15824470
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15824470 1 kg Plastic Bottle 1kg
15814470 500 g Plastic Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15824470 Supplier Supelco Supplier No. 1048641000
 Restricted Item
Beräknad leverans: 18-11-2026
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
For analysis EMSURE™ ACS, ISO, Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7778-50-9
Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.182
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
Synonym potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
PubChem CID 24502
ChEBI CHEBI:53444
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
Specifications
Boiling Point >500°C
Physical Form Crystalline
Quantity 1 kg
Solubility Information Soluble in water: 130g/L at 10°C
Formula Weight 294.19
Percent Purity ≥99.9%
Grade ACS
Odor Odorless
Packaging Plastic Bottle
Density 2.7g/cm3 at 20°C
Chemical Name or Material Potassium dichromate