Läs mer
Potassium Dideuterium Phosphate, CDN
Specifikationer
Specifikationer
| Kemiskt namn eller material | Monopotassium Phosphate-D2 |
| CAS | 13761-79-0 |
| Molekylformel | K . D2 O4 P |
| Kvantitet | 5 g |
| Synonym | Phosphoric acid-d2, potassium salt (1:1) (ACI), Phosphoric acid-d2, monopotassium salt (8 CI, 9 CI), Potassium phosphate (KD2PO4) (6 CI), DKDP, KD*P, KDP-d2, Potassium deuterium phosphate (KD2PO4), Potassium dideuterium phosphate, Potassium dihydrogen phosphate (KD2PO4), Potassium dihydrogen phosphate-d2, Monopotassium phosphate-D2 |
| InChI formel | InChI=1 S/KH3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1/i/hD2 |
| LEDER | [K+].[2 H]OP(=O)([O-])O[2 H] |
| IUPAC-namn | potassium;dideuterio phosphate |
| Molekylvikt (g/mol) | 138.098 |
| Formel vikt | 137.945 g/mol |
| Visa mer |