missing translation for 'onlineSavingsMsg'
Learn More

Potassium Diisopropylxanthate, TRC

Product Code. 30419998
Change view
Click to view available options
Kvantitet:
2.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30419998 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30419998 Supplier Toronto Research Chemicals Supplier No. TRCP6951202.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Potassium Diisopropylxanthate
CAS 3419-34-9
Molekylformel C6H14KO2PS2
Kvantitet 2.5 g
Synonym O,O-Diisopropyl Potassium Phosphorodithioate, Potassium O,O-Diisopropyl Dithiophosphate, Potassium O,O-Diisopropyl Phosphorodithioate, Potassium O,O'-Diisopropyl Dithiophosphate, Potassium Bis(O-isopropyl) Dithiophosphate, Potassium Diisopropyl Dithiophosphate
InChI formel InChI=1S/C6H15O2PS2.K/c1-5(2)7-9(10,11)8-6(3)4;/h5-6H,1-4H3,(H,10,11);/q;+1/p-1
LEDER [K+].CC(C)OP(=S)([S-])OC(C)C
IUPAC-namn potassium;di(propan-2-yloxy)-sulfanylidene-sulfido-λ^{5}-phosphane
Molekylvikt (g/mol) 252.38
Formel vikt 251.981
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.