missing translation for 'onlineSavingsMsg'
Learn More

Potassium ethyltrifluoroborate, 97%, Thermo Scientific Chemicals

Catalog Number p-4602067
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
44248-07-9
C2H5BF3K
135.97
MFCD04112713
GIIPADVFWNGSKY-UHFFFAOYSA-N
potassium ethyltrifluoroborate, potassium ethyltrifluoroboranuide, potassium ethytrifluoroborate, amtb738, ethyltrifluoroborate potassium salt, potassium ethyl trifluoro borate 1-
23668491
potassium;ethyl(trifluoro)boranuide
[K+].CC[B-](F)(F)F
This item is not returnable. View return policy
44248-07-9
C2H5BF3K
135.97
MFCD04112713
GIIPADVFWNGSKY-UHFFFAOYSA-N
potassium ethyltrifluoroborate, potassium ethyltrifluoroboranuide, potassium ethytrifluoroborate, amtb738, ethyltrifluoroborate potassium salt, potassium ethyl trifluoro borate 1-
23668491
potassium;ethyl(trifluoro)boranuide
[K+].CC[B-](F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 80.0°C to 84.0°C
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 135.97
Percent Purity 97%
Chemical Name or Material Potassium ethyltrifluoroborate
compliance-icons
Product Identifier
  • Potassium ethyltrifluoroborate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'