missing translation for 'onlineSavingsMsg'
Learn More

Potassium Ferrocyanide, Trihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number p-5390830
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
14459-95-1
422.39
MFCD00167023
NLYKFUFIUWPPIZ-UHFFFAOYSA-N
tetrapotassium hexacyanoironbis(ylium) trihydrate
O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N
This item is not returnable. View return policy
14459-95-1
422.39
MFCD00167023
NLYKFUFIUWPPIZ-UHFFFAOYSA-N
tetrapotassium hexacyanoironbis(ylium) trihydrate
O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N

High quality chemicals for laboratory and specialized industrial use

Physical Form Crystals
Quantity 500 g
Molecular Formula K4Fe(CN)6·3H2O
Formula Weight 422.41
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.85kg
Chemical Name or Material Potassium Ferrocyanide, Trihydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'