Läs mer
Potassium Hexadecyl Hydrogenphosphate (~90%), TRC
Produktkod. 30414111
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30414111 5 g
1 options
Produktkod. 30414111 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP6951905G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Potassium Hexadecyl Hydrogenphosphate
CAS 19035-79-1
Molekylformel C16H34KO4P
Kvantitet 5 g
Synonym Emulsiphos, Potassium Cetyl Phosphate
InChI formel InChI=1S/C16H35O4P.K/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-20-21(17,18)19;/h2-16H2,1H3,(H2,17,18,19);/q;+1/p-1
LEDER [K+].CCCCCCCCCCCCCCCCOP(=O)(O)[O-]
IUPAC-namn potassium;hexadecyl hydrogen phosphate
Molekylvikt (g/mol) 360.51
Formel vikt 360.1832