Learn More
Potassium hydrogen carbonate, MilliporeSigma™
Product Code. 15894460
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic Bottle
Unit Size:
500g
Product Code. Quantity Packaging Unit Size
15894460 500 g Plastic Bottle 500g
1 options
Product Code. 15894460 Supplier Supelco Supplier No. 1048540500
 Restricted Item
Beräknad leverans: 26-11-2026
For analysis EMSURE™ ACS
TRUSTED_SUSTAINABILITY
Specifications
CAS 298-14-6
Color White
pH 8 to 8.6 (50 g/L at 20°C)
Physical Form Solid
Quantity 500 g
Molecular Formula CHKO3
MDL Number MFCD00011402
Synonym potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
Solubility Information Water: 333g/L at 20°C soluble
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
SMILES [K+].OC([O-])=O
IUPAC Name potassium hydrogen carbonate
Molecular Weight (g/mol) 100.11
PubChem CID 516893
ChEBI CHEBI:81862
Formula Weight 100.12
Percent Purity ≥99.5%
Grade ACS
Odor Odorless
Packaging Plastic Bottle
Density 2.17 g/cm3 (20&de g;C)
Chemical Name or Material Potassium hydrogen carbonate