missing translation for 'onlineSavingsMsg'
Läs mer

Potassium Hydrogen Phthalate, Supelco™

Produktkod. 15824240
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
80 g
Förpackningsstorlek:
80g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15824240 80 g 80g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15824240 Leverantör Supelco Leverantörsnummer 1024000080
  • ONLINE EXCLUSIVE PRICE
    1680.00 SEK
    1292.86 SEK / 80g
    Save 387.14 SEK
    23% Av
Visa alla kampanjer
 Begränsad artikel
I lager
Lägg i varukorgen
Edge
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Volumetric standard, secondary reference material for alkalimetry, traceable to NIST Standard Reference Material (SRM) Certipur™ Reag. Ph Eur

Potassium hydrogen phthalate, also known as KHP, is a volumetric standard manufactured under stringent conditions and measured with the highest possible precision. Its mass fraction is determined by potentiometric titration, and it is qualified in a DIN EN ISO/IEC 17025 accredited laboratory. Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation. In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website. Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

TRUSTED_SUSTAINABILITY

Specifikationer

CAS 877-24-7
Smältpunkt 295°C to 300°C
Densitet 1.636 g/cm3
MDL-nummer MFCD00013070
Kvantitet 80 g
Synonym Potassium hydrogen phthalate, Potassium hydrogen phthalate
Molekylvikt (g/mol) 204.22
Kvalitet Research
Product Identifier
  • Kaliumväteftalat volymstandard, sekundärt referensmaterial för alkalimetri, spårbart till NIST Standard Reference Material (SRM) Certipur® Reag. Ph Eur
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.