Learn More
Potassium Hydrogen Sulfate, Supelco™
Product Code. 15897280
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15897280 2.5 kg 2.5kg
15894470 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15897280 Supplier Supelco Supplier No. 1048852500
 Restricted Item
Beräknad leverans: 26-11-2026
For analysis EMSURE™ Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7646-93-7
Specifications
Melting Point 195°C (decomposition)
Quantity 2.5 kg
MDL Number MFCD00011404
Synonym Potassium hydrogen sulfate, Potassium bisulfate, Potassium hydrogen sulfate
Molecular Weight (g/mol) 136.17
Grade Research
Density 2.24 g/cm3 at 20°C
Chemical Name or Material Potassium Hydrogen Sulfate