Läs mer
Potassium Hydrosulfide, TRC
Produktkod. 30411291
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30411291 100 mg
1 options
Produktkod. 30411291 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP699325100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Potassium Hydrosulfide
CAS 1310-61-8
Molekylformel HKS
Kvantitet 100 mg
Synonym Monopotassium Monohydrogen Sulfide, Potassium Bisulfide, Potassium Hydrogen Sulfide, Potassium Sulfhydrate
InChI formel InChI=1S/K.H2S/h;1H2/q+1;/p-1
LEDER [SH-].[K+]
IUPAC-namn potassium;sulfanide
Molekylvikt (g/mol) 72.17
Formel vikt 71.9436