missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide Solution, volumetric, 0.5M KOH (0.5N), Honeywell Fluka™
Click to view available options
Quantity:
500 mL
2 L
Packaging:
Poly Bottle
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Linear Formula | KOH |
UN Number | UN1814 |
Density | 1.025g/mL at 25°C |
Quantity | 500 mL |
Formula Weight | 56.11g/mol |
Packaging | Poly Bottle |
Physical Form | Liquid |
Chemical Name or Material | Potassium Hydroxide Solution, Volumetric, 0.5M KOH (0.5N) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'