missing translation for 'onlineSavingsMsg'
Learn More

Potassium (iodomethyl)trifluoroborate, tech. 90%, Thermo Scientific Chemicals

Catalog Number 15490749
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
888711-47-5
CH2BF3IK
247.83
MFCD11112187
WQJVWRYIXQXISQ-UHFFFAOYSA-N
potassium trifluoro iodomethyl borate, potassium iodomethyltrifluoroborate, potassium trifluoro iodomethyl boranuide, pubchem11563, potassium iodomethyl trifluoroborate, trifluoro iodomethyl potassioboron v, potassium trifluoro iodomethyl borate 1-, potassium ion trifluoro iodomethyl boranuide, potassium tris fluoranyl-iodanylmethyl boranuide, borate 1-,trifluoro iodomethyl-, potassium, t-4-9ci
23674071
[K+].F[B-](F)(F)CI
This item is not returnable. View return policy
888711-47-5
CH2BF3IK
247.83
MFCD11112187
WQJVWRYIXQXISQ-UHFFFAOYSA-N
potassium trifluoro iodomethyl borate, potassium iodomethyltrifluoroborate, potassium trifluoro iodomethyl boranuide, pubchem11563, potassium iodomethyl trifluoroborate, trifluoro iodomethyl potassioboron v, potassium trifluoro iodomethyl borate 1-, potassium ion trifluoro iodomethyl boranuide, potassium tris fluoranyl-iodanylmethyl boranuide, borate 1-,trifluoro iodomethyl-, potassium, t-4-9ci
23674071
[K+].F[B-](F)(F)CI

Organotrifluoroborate involved in synthesis of functionalized ethyltrifluoroborates, Suzuki-Miyaura cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Organotrifluoroborate involved in synthesis of functionalized ethyltrifluoroborates, Suzuki-Miyaura cross-coupling reactions.

Notes
Light Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
TRUSTED_SUSTAINABILITY
Melting Point 242°C to 258°C
Color Brown to White
Physical Form Solid
Quantity 1 g
Linear Formula ICH2BF3K
IUPAC Name potassium;trifluoro(iodomethyl)boranuide
Formula Weight 247.83
Percent Purity 90%
Grade Technical
Sensitivity Light sensitive
Chemical Name or Material Potassium (iodomethyl)trifluoroborate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'