Läs mer
Potassium Isobutyrate, TRC
Produktkod. 30417937
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30417937 250 mg
1 options
Produktkod. 30417937 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP993420250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Potassium Isobutyrate
CAS 19455-20-0
Molekylformel C4 H7 O2 . K
Kvantitet 250 mg
Synonym Propanoic acid','2-methyl-','potassium salt (1:1) (ACI), Isobutyric acid','potassium salt (8CI), Propanoic acid','2-methyl-','potassium salt (9CI), Potassium 2-methylpropanoate, Potassium isobutyrate, 2-Methylpropanoic acid potassium salt, Isobutyric acid potassium salt
InChI formel InChI=1S/C4H8O2.K/c1-3(2)4(5)6;/h3H,1-2H3,(H,5,6);/q;+1/p-1
LEDER [K+].CC(C)C(=O)[O-]
IUPAC-namn potassium;2-methylpropanoate
Molekylvikt (g/mol) 126.195
Formel vikt 126.008