missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrate, 99%, extra pure, ACROS Organics™

Brand:  Acros Organics 434430100

Additional Details : CAS Number : 7757-79-1 Weight : 10.00000kg Transport : UN number : 1486 Chem class : 5.1 Pack group : III

Packaging Plastic drum
Quantity 10kg

 View more versions of this product

Product Code. 15199293

  • 2072.34 SEK / 10kg


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

7757-79-1
101.1
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
CHEBI:63043
[N+](=O)([O-])[O-].[K+]
KNO3
FGIUAXJPYTZDNR-UHFFFAOYSA-N
24434
potassium;nitrate

Specifications

Potassium nitrate
7757-79-1
KNO3
15,7769
Solubility in water: 320g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol
[N+](=O)([O-])[O-].[K+]
101.1
CHEBI:63043
Fine Crystals or Crystalline Powder
Extra Pure
0.002% max.
0.1% max.
0.001% max.
Solubility in water: 320g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol
400.0°C
334.0°C
Extra Pure
99%
KNO3
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium;nitrate
24434
101.1
99%
0.002% max
10ppm max.
0.1% max. (in water)
Plastic drum
0.005% max
White
10kg
Videos
Documents