Läs mer
Potassium Pentathionate Sesquihydrate, TRC
Produktkod. 17949754
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17949754 100 mg
17948784 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17949754 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP698170100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23371-63-3
Molekylformel 2 K . O6 S5 . 1.5[H2O]
Molekylvikt (g/mol) 354.54916
Synonym Pentathionic acid, dipotassium salt, hydrate (2:3), Pentathionic acid, dipotassium salt, sesquihydrate
LEDER O.[K].[K].OS(=O)(=O)SSSS(=O)(=O)O
Specifikationer
Kemiskt namn eller material Potassium Pentathionate Sesquihydrate
Kvantitet 100 mg
InChI formel InChI=1S/2K.H2O6S5/c;;1-10(2,3)8-7-9-11(4,5)6/h;;(H,1,2,3)(H,4,5,6)/q2*+1;/p-2
Formel vikt 353.7835