missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Permanganate Concentrate, 1/50M, For 1L standard solution, Honeywell Fluka™
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Packaging:
Ampoule
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Color | Violet |
Quantity | 1 Pc. |
UN Number | UN3214 |
Formula Weight | 158.03g/mol |
Concentration or Composition (by Analyte or Components) | 1/50 M KMnO4 (0.1N) |
Packaging | Ampoule |
Appearance | Clear liquid |
Chemical Name or Material | Potassium permanganate concentrate, For 1L standard solution, 1/50M KMnO4 (0.1N) |
Safety and Handling
Product Identifier
- potassium permanganate
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 2
- Skin corrosion/irritation Category 1 B
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : potassium permanganate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'