missing translation for 'onlineSavingsMsg'
Läs mer

Potassium Peroxodisulfate, Supelco™

Produktkod. 15884520
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 g
Förpackningsstorlek:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15884520 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
MQ300 Graded - For Products Used in Applications Requiring Enhanced Change Control and Quality Agreement
Produktkod. 15884520 Leverantör Supelco Leverantörsnummer 1050920250
 Begränsad artikel
Beräknad leverans: 26-11-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy
MQ300 Graded - For Products Used in Applications Requiring Enhanced Change Control and Quality Agreement

For analysis (≤0.001% N) EMSURE™ ACS,Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Potassium Peroxodisulfate
Smältpunkt 100°C (Decomposition)
CAS 7727-21-1
Kvantitet 250 g
MDL-nummer MFCD00011386
Synonym Potassium peroxodisulfate, Potassium persulfate, Potassium peroxodisulfate
Molekylvikt (g/mol) 270.32
Kvalitet Research
Densitet 2.48 g/cm3 at 20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.