missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate, Dibasic, Endotoxin Tested, Multi-Compendial, U.S.P., J.T. Baker™

Catalog Number p-5390850
Click to view available options
Quantity:
500 g
2.5 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
This item is not returnable. View return policy
7758-11-4
HK2O4P
174.17
ZPWVASYFFYYZEW-UHFFFAOYSA-L
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
24450
CHEBI:32031
dipotassium;hydrogen phosphate
OP(=O)([O-])[O-].[K+].[K+]
This item is not returnable. View return policy
7758-11-4
HK2O4P
174.17
ZPWVASYFFYYZEW-UHFFFAOYSA-L
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
24450
CHEBI:32031
dipotassium;hydrogen phosphate
OP(=O)([O-])[O-].[K+].[K+]
Quantity 500 g
Formula Weight 174.18
Grade Multi-Compendial/USP
Packaging Poly Bottle
Chemical Name or Material Potassium Phosphate, Dibasic, Endotoxin Tested
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'