Läs mer
Potassium propionate, TRC
Produktkod. 30412699
Change view
Kvantitet:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30412699 50 g
30427274 5 g
30418934 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30412699 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP69529350G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 327-62-8
Molekylformel C3H5KO2
Molekylvikt (g/mol) 112.17
Synonym Propanoic acid, potassium salt (9CI), Propionic acid, potassium salt (8CI), Potassium propanoate, Potassium propionate
IUPAC-namn potassium;propanoate
LEDER [K+].CCC(=O)[O-]
Specifikationer
Kemiskt namn eller material Potassium propionate
Kvantitet 50 g
InChI formel InChI=1S/C3H6O2.K/c1-2-3(4)5;/h2H2,1H3,(H,4,5);/q;+1/p-1
Formel vikt 111.9927