missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Rhodizonate, MP Biomedicals
Click to view available options
Quantity:
1 g
Specifications
CAS | 13021-40-4 |
Molecular Formula | C6H2KO6 |
MDL Number | MFCD00001649 |
Quantity | 1 g |
Synonym | potassium rhodizonate, rhodizonic acid dipotassium salt, dipotassium rhodizonate, 5-cyclohexene-1,2,3,4-tetrone, 5,6-dihydroxy-, dipotassium salt, potassium 3,4,5,6-tetraoxocyclohex-1-ene-1,2-bis olate, 5-cyclohexene-1,2,3,4-tetrone, 5,6-dihydroxy-, potassium salt 1:2, dipotassium 3,4,5,6-tetraoxocyclohexene-1,2-diolate, kaliumrhodizonat, dipotassium tetraoxocyclohex-1-ene-1,2-bis olate, potassium rhodizonate, technical grade |
InChI Key | UGFHFZSZCJFOTO-UHFFFAOYSA-N |
SMILES | [K].OC1=C(O)C(=O)C(=O)C(=O)C1=O |
IUPAC Name | dihydroxycyclohex-5-ene-1,2,3,4-tetrone potassium |
Molecular Weight (g/mol) | 209.17 |
PubChem CID | 83051 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'