missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Sodium Tartrate, 4-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
Specifications
Physical Form | Crystals |
Quantity | 1 kg |
Formula Weight | 282.22 |
Grade | BAKER ANALYZED™ ACS Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 1.79kg |
Chemical Name or Material | Potassium Sodium Tartrate, 4-Hydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'