Learn More
Potassium Sulfate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Catalog Number 15528114
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
High quality chemicals for laboratory and specialized industrial use
CAS 7778-80-5
Physical Form Powder
Quantity 500 g
Molecular Formula K2O4S
MDL Number MFCD00011388
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
SMILES [K+].[K+].[O-]S([O-])(=O)=O
IUPAC Name dipotassium sulfate
Molecular Weight (g/mol) 174.25
PubChem CID 24507
ChEBI CHEBI:32036
Formula Weight 174.27
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.66kg
Chemical Name or Material Potassium Sulfate