Läs mer
Kaliumtetrakloroplatinum(II), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Potassium Tetrachloroplatinum(II) |
| CAS | 10025-99-7 |
| Molekylformel | Cl4 Pt . 2 K |
| Kvantitet | 250 mg |
| Synonym | Platinate(2-)','tetrachloro-','potassium (1:2)','(SP-4-1)-, Platinate(2-)','tetrachloro-','dipotassium (8CI), Platinate(2-)','tetrachloro-','dipotassium','(SP-4-1)- (9CI), Potassium chloroplatinate(II) (6CI), Dipotassium platinum tetrachloride, Dipotassium tetrachloroplatinate, Dipotassium tetrachloroplatinate(2-), Dipotassium tetrachloroplatinate(II), NSC 175496, Platinous potassium chloride, Platinum potassium chloride (PtK2Cl4), Potassium chloroplatinate, Potassium chloroplatinate (K2PtCl4), Potassium platinochloride, Potassium platinous chloride, Potassium platinum(II) tetrachloride, Potassium tetrachloroplatinate, Potassium tetrachloroplatinate K2PtCl4, Potassium tetrachloroplatinate(2-), Potassium tetrachloroplatinate(II), Potassium tetrachloroplatinum(II), Tetrachloroplatinum(II) acid potassium salt |
| InChI formel | InChI=1S/4ClH.2K.Pt/h4*1H;;;/q;;;;2*+1;+2/p-4 |
| LEDER | [K+].[K+].[Cl-][Pt+2]([Cl-])([Cl-])[Cl-] |
| Molekylvikt (g/mol) | 415.09 |
| Formel vikt | 412.7676 |