missing translation for 'onlineSavingsMsg'
Learn More

Potassium Thiocyanate, Certified AR for Analysis, Fisher Chemical™

Catalog Number p-7073453
Click to view available options
Quantity:
250 g
500 g
333-20-0
CKNS
97.176
11413
ZNNZYHKDIALBAK-UHFFFAOYSA-M
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
516872
CHEBI:30951
potassium;thiocyanate
C(#N)[S-].[K+]
333-20-0
CKNS
97.176
11413
ZNNZYHKDIALBAK-UHFFFAOYSA-M
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
516872
CHEBI:30951
potassium;thiocyanate
C(#N)[S-].[K+]
Melting Point 170°C
Color Colorless to White
pH 5.3 to 8.7
Physical Form Crystalline Solid
Quantity 250 g
Formula Weight 97.18g/mol
Packaging Plastic powder jar
Vapor Pressure <1 hPa at 20°C
compliance-icons
Product Identifier
  • Potassium thiocyanate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P233-Keep container tightly closed.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'