Läs mer
Potassium Trichloroammineplatinate, TRC
Produktkod. 30421284
Change view
Kvantitet:
10 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30421284 100 mg
30423387 10 mg
30425146 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30421284 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP698550100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13820-91-2
Molekylformel K . Cl3 H3 N Pt
Molekylvikt (g/mol) 357.57
Synonym Platinate(1-), amminetrichloro-, potassium, (SP-4-2)- (9CI), Platinate(1-), amminetrichloro-, potassium (8CI), Potassium trichloroammineplatinate(II) (6CI,7CI), NSC 123898, Potassium amminetrichloroplatinate, Potassium amminetrichloroplatinate(1-) Coordination Compound
LEDER [K+].[Cl-][Pt+2]([Cl-])([Cl-])[NH3]
Specifikationer
Kemiskt namn eller material Potassium Trichloroammineplatinate
Kvantitet 100 mg
InChI formel InChI=1S/3ClH.K.H3N.Pt/h3*1H;;1H3;/q;;;+1;;+2/p-3
Formel vikt 355.8616