Learn More
Potassium trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals
Description
Potassium trifluoromethanesulfonate is used as a reagent in the synthesis of guanine-quadruplex hybrid materials. It acts as a supporting electrolyte in the electrochemical study of evidence for gold anion in ethylenediamine. It is involved in the preparation of N-fluoro-2,4,6-trimethylpyridinium triflate by reacting with 2,4,6-trimethyl-pyridine. Further, it is used in the preparation of imidazolium salt, 3-methyl-1-(3R,3aR,6S,6aR)-[6-(benzyloxy)-hexahydrofuro[3,2-b]furan-3-yl]imidazolium trifluoromethanesulfonate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 10 g |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 188.17 |
Percent Purity | 98% |
Physical Form | Crystalline |
Chemical Name or Material | Potassium trifluoromethanesulfonate |
RUO – Research Use Only