Läs mer
Potassiummethyldithiocarbamate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-Methyldithiocarbamate Potassium |
| CAS | 137-41-7 |
| Molekylformel | C2 H4 N S2 . K |
| Kvantitet | 250 mg |
| Synonym | Potassiummethyldithiocarbamate, Carbamic acid','methyldithio-','monopotassium salt (8CI), Carbamic acid','methyldithio-','potassium salt (6CI), Dicid, K-Pam HL, Metam potassium, Metam-potassium, Potassium methyldithiocarbamate, Potassium N-methyldithiocarbamate, N-Methyldithiocarbamate Pota |
| InChI formel | InChI=1S/C2H5NS2.K/c1-3-2(4)5;/h1H3,(H2,3,4,5);/q;+1/p-1 |
| LEDER | [K+].CNC(=S)[S-] |
| IUPAC-namn | potassium;N-methylcarbamodithioate |
| Molekylvikt (g/mol) | 145.288 |
| Formel vikt | 144.942 |