Learn More
Pouds™ Wall Brackets Product Code.: 12993505
Pouds™ Wall Brackets
Product Code. 12993505
Sharpsguard mini, 1, 2.5, 3.75, 5 and com plus, Each
Click to view available options
For Use With (Equipment):
Sharpsguard 7, 11.5, and 22
Sharpsguard mini, 1, 2.5, 3.75, 5 and com plus
Unit Size:
Each
Product Code. 12993505
Brand: Daniels Healthcare DD487
For attaching sharps containers to the wall
Wall bracket for SHARPSGUARD™ mini, 1, 2.5, 3.75, 5 and com plus containers. Red.
Specifications
Color Red
For Use With (Application) Attaching sharps container to a wall
For Use With (Equipment) Sharpsguard mini, 1, 2.5, 3.75, 5 and com plus
Product Content Correction
Product Title
Pouds™ Wall Brackets >