Learn More
WLD Tec™ Powerjet 2 Buckling Gun
Product Code. 10367492
Change view
Click to view available options
Height (Metric):
210 mm
Unit Size:
Each
Product Code. Height (Metric) Unit Size
10367492 210 mm Each
1 options
Product Code. 10367492 Supplier WLD Tec™ Supplier No. 6.006.000
  • ONLINE EXCLUSIVE PRICE
    7864.21 SEK
    5330.28 SEK / Paket med 1
    Save 2533.93 SEK
    32% Av
Beräknad leverans: 17-12-2026
Specifications
Product Type Laboratory Gas Burner
Gas Type Liquid Gas, Natural Gas
Length (Metric) 150 mm
For Use With (Application) Microbiology or Biotechnical Fields
Height (Metric) 210 mm
Max. Temperature (Metric) 1300°C (Natural Gas), 1350°C (Liquid Gas)
Weight (Metric) 500 g
Width (Metric) 150 mm